3-Acetamidopyridine

Supplier: Matrix Scientific

077210-500MG 077210-1G
102665-058EA 161.54 USD
102665-058 102665-060
3-Acetamidopyridine
3-Acetamidopyridine

Hazard: Irritant

Formula: C₇H₈N₂O
MW: 136.15 g/mol
CAS Number: 5867-45-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR