Cyclohexanone oxime

Supplier: Matrix Scientific

077206-500MG 077206-1G 077206-5G
102665-046EA 161.54 USD
102665-046 102665-048 102665-050
Cyclohexanone oxime
Cyclohexanone oxime

Hazard: Irritant

Formula: C₆H₁₁NO
MDL Number: MFCD00001660
CAS Number: 100-64-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR