3-(Chloromethyl)-1-methylpyrrolidine hydrochloride

Supplier: Matrix Scientific

077203-500MG 077203-1G 077203-5G
102665-028EA 692.31 USD
102665-028 102665-030 102665-032
3-(Chloromethyl)-1-methylpyrrolidine hydrochloride
3-(Chloromethyl)-1-methylpyrrolidine hydrochloride

Hazard: Irritant

Formula: C₆H₁₂ClN·HCl
MW: 170.08 g/mol
MDL Number: MFCD20441425
CAS Number: 56669-84-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR