2-(2-Chloroethyl)-1H-benzimidazole hydrochloride
Supplier: Matrix Scientific
077201-500MG
077201-1G
102665-018EA
516.92
USD
102665-018
102665-020
2-(2-Chloroethyl)-1H-benzimidazole hydrochloride
2-(2-Chloroethyl)-1H-benzimidazole hydrochloride
Hazard: Irritant
Formula:
C₉H₁₀Cl₂N₂ MW: 217.1 g/mol |
MDL Number:
MFCD20441422 CAS Number: 405173-97-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...