1-(Ethylamino)propan-2-ol

Supplier: Matrix Scientific

077200-500MG 077200-1G 077200-5G
102665-012EA 516.92 USD
102665-012 102665-014 102665-016
1-(Ethylamino)propan-2-ol
1-(Ethylamino)propan-2-ol

Hazard: Irritant

Formula: C₅H₁₃NO
MDL Number: MFCD00082193
CAS Number: 40171-86-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR