2-Nitro-1-propanol
Supplier: Matrix Scientific
Synonyms:
1-Hydroxy-2-nitropropane
077199-500MG
077199-1G
077199-5G
102665-006EA
161.54
USD
102665-006
102665-008
102665-010
2-Nitro-1-propanol
2-Nitro-1-propanol
Hazard: Irritant
Formula:
C₃H₇NO₃ MW: 105.09 g/mol |
MDL Number:
MFCD00007398 CAS Number: 2902-96-7 |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...