3,5-Diamino-1H-1,2,4-triazole
Supplier: Matrix Scientific
Synonyms:
3,5-Diamino-1,2,4-triazole, Guanazole
077198-500MG
077198-1G
077198-5G
102665-000EA
161.54
USD
102665-000
102665-002
102665-004
3,5-Diamino-1H-1,2,4-triazole
3,5-Diamino-1H-1,2,4-triazole
Hazard: Irritant
Formula:
C₂H₅N₅ MW: 99.1 g/mol |
MDL Number:
MFCD00005233 CAS Number: 1455-77-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...