2-{[(4-Ethoxyphenyl)amino]methyl}-1H-isoindole-1,3(2H)-dione

Supplier: Matrix Scientific

077165-500MG 077165-1G 077165-5G
102664-818EA 258.46 USD
102664-818 102664-820 102664-822
2-{[(4-Ethoxyphenyl)amino]methyl}-1H-isoindole-1,3(2H)-dione
2-{[(4-Ethoxyphenyl)amino]methyl}-1H-isoindole-1,3(2H)-dione

Hazard: Irritant

Formula: C₁₇H₁₆N₂O₃
MDL Number: MFCD00087142
CAS Number: 7506-36-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR