4-[(2-Methylphenyl)amino]-4-oxobutanoic acid

Supplier: Matrix Scientific

077135-500MG 077135-1G 077135-5G
102664-642EA 161.54 USD
102664-642 102664-644 102664-646
4-[(2-Methylphenyl)amino]-4-oxobutanoic acid
4-[(2-Methylphenyl)amino]-4-oxobutanoic acid

Hazard: Irritant

Formula: C₁₁H₁₃NO₃
MDL Number: MFCD00089587
CAS Number: 37600-45-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR