4-[(4-Chlorophenyl)amino]-4-oxobutanoic acid

Supplier: Matrix Scientific

077121-500MG 077121-1G 077121-5G
102664-558EA 161.54 USD
102664-558 102664-560 102664-562
4-[(4-Chlorophenyl)amino]-4-oxobutanoic acid
4-[(4-Chlorophenyl)amino]-4-oxobutanoic acid

Hazard: Irritant

Formula: C₁₀H₁₀ClNO₃
MDL Number: MFCD00029843
CAS Number: 17722-52-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR