4-[(3,4-Dimethylphenyl)amino]-4-oxobutanoic acid

Supplier: Matrix Scientific

077119-500MG 077119-1G
102664-548EA 161.54 USD
102664-548 102664-550
4-[(3,4-Dimethylphenyl)amino]-4-oxobutanoic acid
4-[(3,4-Dimethylphenyl)amino]-4-oxobutanoic acid

Hazard: Irritant

Formula: C₁₂H₁₅NO₃
MDL Number: MFCD00029836
CAS Number: 69517-61-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR