2-(3,4-Dimethylphenyl)-1,3-dioxoisoindoline-5-carboxylic acid

Supplier: Matrix Scientific

077118-500MG 077118-1G 077118-5G
102664-542EA 193.85 USD
102664-542 102664-544 102664-546
2-(3,4-Dimethylphenyl)-1,3-dioxoisoindoline-5-carboxylic acid
2-(3,4-Dimethylphenyl)-1,3-dioxoisoindoline-5-carboxylic acid

Hazard: Irritant

Formula: C₁₇H₁₃NO₄
MDL Number: MFCD00404561
CAS Number: 294667-04-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR