2-(3-Chloro-4-methylphenyl)-1,3-dioxoisoindoline-5-carboxylic acid

Supplier: Matrix Scientific

077117-500MG 077117-1G 077117-5G
102664-536EA 193.85 USD
102664-536 102664-538 102664-540
2-(3-Chloro-4-methylphenyl)-1,3-dioxoisoindoline-5-carboxylic acid
2-(3-Chloro-4-methylphenyl)-1,3-dioxoisoindoline-5-carboxylic acid

Hazard: Irritant

Formula: C₁₆H₁₀ClNO₄
MDL Number: MFCD00981453
CAS Number: 313260-37-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR