[4-(Aminomethyl)cyclohexyl]methanol

Supplier: Matrix Scientific

077092-500MG 077092-1G 077092-5G
102664-432EA 455.38 USD
102664-432 102664-434 102664-436
[4-(Aminomethyl)cyclohexyl]methanol
[4-(Aminomethyl)cyclohexyl]methanol

Hazard: Irritant

Formula: C₈H₁₇NO
MDL Number: MFCD00152235
CAS Number: 1074-62-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR