N-Benzyltetrahydro-2H-pyran-4-amine hydrochloride

Supplier: Matrix Scientific

Synonyms: Benzyl-(tetrahydro-pyran-4-yl)-amine

077091-500MG 077091-1G
102664-428EA 516.92 USD
102664-428 102664-430
N-Benzyltetrahydro-2H-pyran-4-amine hydrochloride
N-Benzyltetrahydro-2H-pyran-4-amine hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₈ClNO
MW: 227.74 g/mol
MDL Number: MFCD11035934
CAS Number: 443344-23-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR