(1-Methyl-1H-pyrazol-5-yl)methanol
Supplier: Matrix Scientific
Synonyms:
5-(Hydroxymethyl)-1-methylpyrazole, 1-Methylpyrazole-5-methanol
077089-1G
077089-5G
102664-420EA
90.77
USD
102664-420
102664-422
(1-Methyl-1H-pyrazol-5-yl)methanol
(1-Methyl-1H-pyrazol-5-yl)methanol
Hazard: Irritant
Formula:
C₅H₈N₂O MW: 112.13 g/mol |
MDL Number:
MFCD08556301 CAS Number: 84547-61-5 |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...