N-(Cyclopropylmethyl)ethanamine

Supplier: Matrix Scientific

077086-500MG 077086-1G 077086-5G
102664-408EA 516.92 USD
102664-408 102664-410 102664-412
N-(Cyclopropylmethyl)ethanamine
N-(Cyclopropylmethyl)ethanamine

Hazard: Irritant

Formula: C₆H₁₃N
MDL Number: MFCD11140436
CAS Number: 26389-65-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR