Methyl-4-bromo-1H-indole-2-carboxylate

Supplier: Matrix Scientific

Synonyms: Methyl 4-bromo-1H-indole-2-carboxylate

077082-500MG 077082-1G 077082-5G
102664-392EA 387.69 USD
102664-392 102664-394 102664-396
Methyl-4-bromo-1H-indole-2-carboxylate
Methyl-4-bromo-1H-indole-2-carboxylate

Hazard: Irritant

Formula: C₁₀H₈BrNO₂
MW: 254.09 g/mol
MDL Number: MFCD04966956
CAS Number: 167479-13-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR