4-(2-Fluorobenzyloxy)benzaldehyde
Supplier: Matrix Scientific
Synonyms:
4-((2-Fluorobenzyl)oxy)benzaldehyde
077076-500MG
077076-1G
077076-5G
102664-382EA
258.46
USD
102664-382
102664-384
102664-386
4-(2-Fluorobenzyloxy)benzaldehyde
4-(2-Fluorobenzyloxy)benzaldehyde
Hazard: Irritant
Formula:
C₁₄H₁₁FO₂ MW: 230.234 g/mol |
MDL Number:
MFCD01590568 CAS Number: 70627-20-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...