4-Fluoro-1H-pyrazole ≥95%

Supplier: Matrix Scientific

077061-250MG 077061-1G
102833-934EA 176.92 USD
102833-934 102833-936
4-Fluoro-1H-pyrazole ≥95%
4-Fluoro-1H-pyrazole

Hazard: Irritant/Refrigerate

Formula: C₃H₃FN₂
Boiling Pt: 72…74 °C (9 mmHg)
Melting Pt: 34…38 °C
MDL Number: MFCD10699125
CAS Number: 35277-02-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR