3-Bromo-4-(trifluoromethyl)benzyl alcohol ≥98%

Supplier: Matrix Scientific

Synonyms: Benzenemethanol, 3-bromo-4-(trifluoromethyl)

077059-250MG 077059-1G
102664-350EA 122.92 USD
102664-350 102664-352
3-Bromo-4-(trifluoromethyl)benzyl alcohol ≥98%
3-Bromo-4-(trifluoromethyl)benzyl alcohol

Hazard: Irritant

Formula: C₈H₆BrF₃O
MDL Number: MFCD13185397
CAS Number: 372120-54-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR