tert-Butyl-N-(2-oxo-2,3,4,5-tetrahydro-1H-1-benzazepin-3-yl)carbamate ≥97%

Supplier: Matrix Scientific

077054-500MG 077054-1G
102664-338EA 600 USD
102664-338 102664-340
tert-Butyl-N-(2-oxo-2,3,4,5-tetrahydro-1H-1-benzazepin-3-yl)carbamate ≥97%
tert-Butyl-N-(2-oxo-2,3,4,5-tetrahydro-1H-1-benzazepin-3-yl)carbamate

Hazard: Irritant

Formula: C₁₅H₂₀N₂O₃
MW: 276.34 g/mol
MDL Number: MFCD19441845
CAS Number: 86499-69-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR