Methyl-4-hydroxy-2,2-dimethyl-2,3-dihydro-1-benzofuran-6-carboxylate ≥95%

Supplier: Matrix Scientific

077044-500MG 077044-1G
102664-304EA 600 USD
102664-304 102664-306
Methyl-4-hydroxy-2,2-dimethyl-2,3-dihydro-1-benzofuran-6-carboxylate ≥95%
Methyl-4-hydroxy-2,2-dimethyl-2,3-dihydro-1-benzofuran-6-carboxylate

Hazard: Irritant

Formula: C₁₂H₁₄O₄
MDL Number: MFCD11226227
CAS Number: 955884-97-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR