Methyl-5-acetyl-4-bromo-1H-pyrrole-2-carboxylate ≥97%

Supplier: Matrix Scientific

077040-500MG 077040-1G
102664-290EA 403.08 USD
102664-290 102664-292
Methyl-5-acetyl-4-bromo-1H-pyrrole-2-carboxylate ≥97%
Methyl-5-acetyl-4-bromo-1H-pyrrole-2-carboxylate

Hazard: Irritant

Formula: C₈H₈BrNO₃
MW: 246.07 g/mol
Melting Pt: 123…125 °C
MDL Number: MFCD22383940
CAS Number: 157425-54-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR