3-Iodo-8-nitroquinoline ≥95%

Supplier: Matrix Scientific

077037-500MG 077037-1G
102664-278EA 298.46 USD
102664-278 102664-280
3-Iodo-8-nitroquinoline ≥95%
3-Iodo-8-nitroquinoline

Hazard: Irritant

Formula: C₉H₅IN₂O₂
Melting Pt: 120…121 °C
MDL Number: MFCD09261162
CAS Number: 497084-46-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR