4-Iodo-1,2-dihydropyridin-2-one ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Iodopyridin-2(1H)-one, 4-Iodo-1H-pyridin-2-one

077035-500MG 077035-1G
102664-270EA 392.31 USD
102664-270 102664-272
4-Iodo-1,2-dihydropyridin-2-one ≥95%
4-Iodo-1,2-dihydropyridin-2-one

Hazard: Irritant

Formula: C₅H₄INO
MW: 221 g/mol
MDL Number: MFCD09702419
CAS Number: 858839-90-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR