Diethyl-2-{[(6-methoxypyridin-3-yl)amino]methylene}malonate ≥97%

Supplier: Matrix Scientific

Synonyms: DIethyl 2-{[(6-methoxypyridin-3-yl)amino]methylene}malonate

077034-500MG 077034-1G 077034-5G
102664-264EA 193.85 USD
102664-264 102664-266 102664-268
Diethyl-2-{[(6-methoxypyridin-3-yl)amino]methylene}malonate ≥97%
Diethyl-2-{[(6-methoxypyridin-3-yl)amino]methylene}malonate

Hazard: Irritant

Formula: C₁₄H₁₈N₂O₅
MW: 294.31 g/mol
MDL Number: MFCD08457989
CAS Number: 53241-90-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR