Ethyl-2-(4,6-dihydroxypyrimidin-5-yl)acetate ≥97%

Supplier: Matrix Scientific

077031-500MG 077031-1G
102664-252EA 600 USD
102664-252 102664-254
Ethyl-2-(4,6-dihydroxypyrimidin-5-yl)acetate ≥97%
Ethyl-2-(4,6-dihydroxypyrimidin-5-yl)acetate

Hazard: Irritant

Formula: C₈H₁₀N₂O₄
MW: 198.18 g/mol
MDL Number: MFCD20527459
CAS Number: 1095822-20-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR