6,8-Dichloro-3-formylchromone ≥95%

Supplier: Matrix Scientific

Synonyms: 6,8-Dichloro-3-formylchromone, 6,8-Dichlorochromone-3-carboxaldehyde, 6,8-Dichloro-4-oxo-4H-chromene-3-carbaldehyde, 6,8-Dichloro-4-oxo-4H-1-benzopyran-3-carboxaldehyde

077019-500MG 077019-1G
102664-206EA 193.85 USD
102664-206 102664-208
6,8-Dichloro-3-formylchromone ≥95%
6,8-Dichloro-3-formylchromone

Hazard: Irritant

Formula: C₁₀H₄Cl₂O₃
MW: 243.0430 g/mol
MDL Number: MFCD00051510
CAS Number: 64481-10-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR