2,6-Diamino-5-chloropyrimidin-4-ol ≥97%

Supplier: Matrix Scientific

077018-500MG 077018-1G
102664-202EA 273.85 USD
102664-202 102664-204
2,6-Diamino-5-chloropyrimidin-4-ol ≥97%
2,6-Diamino-5-chloropyrimidin-4-ol

Hazard: Irritant

Formula: C₄H₅ClN₄O
Melting Pt: 316…318 °C
MDL Number: MFCD22196157
CAS Number: 71552-22-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR