N-Cyclopropyl-2-nitroaniline ≥97%

Supplier: Matrix Scientific

077016-500MG 077016-1G
102664-192EA 298.46 USD
102664-192 102664-194
N-Cyclopropyl-2-nitroaniline ≥97%
N-Cyclopropyl-2-nitroaniline

Hazard: Irritant

Formula: C₉H₁₀N₂O₂
MDL Number: MFCD06149982
CAS Number: 55432-23-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR