8-Chloro-6-(trifluoromethyl)[1,2,4]triazolo[1,5-a]pyridin-2-amine ≥95%

Supplier: Matrix Scientific

077012-500MG 077012-1G
102664-176EA 403.08 USD
102664-176 102664-178
8-Chloro-6-(trifluoromethyl)[1,2,4]triazolo[1,5-a]pyridin-2-amine ≥95%
8-Chloro-6-(trifluoromethyl)[1,2,4]triazolo[1,5-a]pyridin-2-amine

Hazard: Irritant

Formula: C₇H₄ClF₃N₄
MW: 236.58 g/mol
MDL Number: MFCD22383935
CAS Number: 1206640-61-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR