6-Bromo-1-cyclopentyl-1H,2H,3H-imidazo[4,5-b]pyridin-2-one ≥97%

Supplier: Matrix Scientific

077004-500MG 077004-1G
102664-146EA 403.08 USD
102664-146 102664-148
6-Bromo-1-cyclopentyl-1H,2H,3H-imidazo[4,5-b]pyridin-2-one ≥97%
6-Bromo-1-cyclopentyl-1H,2H,3H-imidazo[4,5-b]pyridin-2-one

Hazard: Irritant

Formula: C₁₁H₁₂BrN₃O
Melting Pt: 165…168 °C
MDL Number: MFCD22383941
CAS Number: 1393845-82-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR