4,5-Bis(2,2-dimethyl-1,3-dioxolan-4-yl)-2,2-dimethyl-1,3-dioxolane ≥95%

Supplier: Matrix Scientific

077002-500MG 077002-1G 077002-5G
102664-136EA 193.85 USD
102664-136 102664-138 102664-140
4,5-Bis(2,2-dimethyl-1,3-dioxolan-4-yl)-2,2-dimethyl-1,3-dioxolane ≥95%
4,5-Bis(2,2-dimethyl-1,3-dioxolan-4-yl)-2,2-dimethyl-1,3-dioxolane

Hazard: Irritant

Formula: C₁₅H₂₆O₆
MDL Number: MFCD00138009
CAS Number: 1214702-92-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR