O-[(Benzyloxy)methyl]hydroxylamine ≥97%

Supplier: Matrix Scientific

077001-500MG 077001-1G
102664-132EA 403.08 USD
102664-132 102664-134
O-[(Benzyloxy)methyl]hydroxylamine ≥97%
O-[(Benzyloxy)methyl]hydroxylamine

Hazard: Irritant

Formula: C₈H₁₁NO₂
MDL Number: MFCD11111598
CAS Number: 895588-70-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR