(2S)-3-Amino-2-methylpropan-1-ol ≥95%

Supplier: Matrix Scientific

Synonyms: (S)-3-Amino-2-methylpropan-1-ol

077000-500MG 077000-1G
102664-128EA 821.54 USD
102664-128 102664-130
(2S)-3-Amino-2-methylpropan-1-ol ≥95%
(2S)-3-Amino-2-methylpropan-1-ol

Hazard: Irritant

Formula: C₄H₁₁NO
MW: 89.14 g/mol
MDL Number: MFCD19203687
CAS Number: 88586-62-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR