2-Amino-N-methyl-2-phenylacetamide ≥90%

Supplier: Matrix Scientific

076999-500MG 076999-1G
102664-124EA 323.08 USD
102664-124 102664-126
2-Amino-N-methyl-2-phenylacetamide ≥90%
2-Amino-N-methyl-2-phenylacetamide

Hazard: Irritant

Formula: C₉H₁₂N₂O
MDL Number: MFCD09948081
CAS Number: 93782-07-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR