5-Amino-4-methyl-3,4-dihydro-2H-1,4-benzoxazin-3-one ≥95%

Supplier: Matrix Scientific

076998-500MG 076998-1G
102664-120EA 403.08 USD
102664-120 102664-122
5-Amino-4-methyl-3,4-dihydro-2H-1,4-benzoxazin-3-one ≥95%
5-Amino-4-methyl-3,4-dihydro-2H-1,4-benzoxazin-3-one

Hazard: Irritant

Formula: C₉H₁₀N₂O₂
Melting Pt: 109…111 °C
MDL Number: MFCD12406817
CAS Number: 132522-83-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR