(2S)-2-Amino-3-methylbutan-1-ol hydrochloride ≥95%

Supplier: Matrix Scientific

076997-500MG 076997-1G
102664-116EA 403.08 USD
102664-116 102664-118
(2S)-2-Amino-3-methylbutan-1-ol hydrochloride ≥95%
(2S)-2-Amino-3-methylbutan-1-ol hydrochloride

Hazard: Irritant

Formula: C₅H₁₄ClNO
MDL Number: MFCD00050559
CAS Number: 17016-89-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR