2-(2-Phenylacetamido)benzoic acid ≥97%

Supplier: Matrix Scientific

076657-500MG 076657-1G
102547-308EA 298.46 USD
102547-308 102547-310
2-(2-Phenylacetamido)benzoic acid ≥97%
2-(2-Phenylacetamido)benzoic acid

Hazard: Irritant

Formula: C₁₅H₁₃NO₃
MDL Number: MFCD00437388
CAS Number: 28565-98-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR