2-(10H-Phenothiazin-10-yl)ethan-1-amine hydrochloride ≥97%

Supplier: Matrix Scientific

076656-500MG 076656-1G
102547-304EA 403.08 USD
102547-304 102547-306
2-(10H-Phenothiazin-10-yl)ethan-1-amine hydrochloride ≥97%
2-(10H-Phenothiazin-10-yl)ethan-1-amine hydrochloride

Hazard: Irritant

Formula: C₁₄H₁₅ClN₂S
MDL Number: MFCD22192242
CAS Number: 50971-79-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR