5-Nitro-3,4-dihydro-2H-1,4-benzoxazine ≥97%

Supplier: Matrix Scientific

076655-500MG 076655-1G 076655-5G
102547-298EA 343.08 USD
102547-298 102547-300 102547-302
5-Nitro-3,4-dihydro-2H-1,4-benzoxazine ≥97%
5-Nitro-3,4-dihydro-2H-1,4-benzoxazine

Hazard: Irritant

Formula: C₈H₈N₂O₃
Melting Pt: 96…98 °C
MDL Number: MFCD09955629
CAS Number: 137469-90-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR