Methyl (2S)-1-(3,3-dimethyl-2-oxopentanoyl)pyrrolidine-2-carboxylate ≥97%

Supplier: Matrix Scientific

076645-500MG 076645-1G
102547-254EA 803.08 USD
102547-254 102547-256
Methyl (2S)-1-(3,3-dimethyl-2-oxopentanoyl)pyrrolidine-2-carboxylate ≥97%
Methyl (2S)-1-(3,3-dimethyl-2-oxopentanoyl)pyrrolidine-2-carboxylate

Hazard: Irritant

Formula: C₁₃H₂₁NO₄
MW: 255.32 g/mol
MDL Number: MFCD02179273
CAS Number: 186268-77-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR