Methyl-2-(2-amino-5-ethyl-1,3-thiazol-4-yl)acetate ≥97%

Supplier: Matrix Scientific

076642-500MG 076642-1G 076642-5G
102547-240EA 298.46 USD
102547-240 102547-242 102547-244
Methyl-2-(2-amino-5-ethyl-1,3-thiazol-4-yl)acetate ≥97%
Methyl-2-(2-amino-5-ethyl-1,3-thiazol-4-yl)acetate

Hazard: Irritant

Formula: C₈H₁₂N₂O₂S
Melting Pt: 131…133 °C
MDL Number: MFCD22373664
CAS Number: 1379610-44-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR