Ethyl-4,5,6,7-tetrahydro-1H-indazole-3-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 4,5,6,7-tetrahydro-1H-indazole-3-carboxylate

076636-500MG 076636-1G 076636-5G
102547-212EA 238.46 USD
102547-212 102547-214 102547-216
Ethyl-4,5,6,7-tetrahydro-1H-indazole-3-carboxylate ≥95%
Ethyl-4,5,6,7-tetrahydro-1H-indazole-3-carboxylate

Hazard: Irritant

Formula: C₁₀H₁₄N₂O₂
MW: 194.24 g/mol
Melting Pt: 88…90 °C
MDL Number: MFCD02942367
CAS Number: 4492-02-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR