Ethyl-1H-pyrrolo[2,3-c]pyridine-7-carboxylate ≥97%

Supplier: Matrix Scientific

076635-500MG 076635-1G
102547-208EA 1192.31 USD
102547-208 102547-210
Ethyl-1H-pyrrolo[2,3-c]pyridine-7-carboxylate ≥97%
Ethyl-1H-pyrrolo[2,3-c]pyridine-7-carboxylate

Hazard: Irritant

Formula: C₁₀H₁₀N₂O₂
MW: 190.2 g/mol
Melting Pt: 112…114 °C
MDL Number: MFCD20685899
CAS Number: 945840-74-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR