Ethyl-4-{2,4-dioxo-3-azatricyclo[7.3.1.0

Supplier: Matrix Scientific

Synonyms: Ethyl 4-(1,3-dioxo-1H-benzo[de]isoquinolin-2(3H)-yl)butanoate

076632-500MG 076632-1G 076632-5G
102547-192EA 298.46 USD
102547-192 102547-194 102547-196
Ethyl-4-{2,4-dioxo-3-azatricyclo[7.3.1.0
Ethyl-4-{2,4-dioxo-3-azatricyclo[7.3.1.0^{5,13}]trideca-1(13),5,7,9,11-pentaen-3-yl}butanoate

Hazard: Irritant

Formula: C₁₈H₁₇NO₄
MW: 311.34 g/mol
Melting Pt: 91…92 °C
MDL Number: MFCD00392161
CAS Number: 150705-10-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR