(2S)-1-(3,3-Dimethyl-2-oxopentanoyl)pyrrolidine-2-carboxylic acid ≥97%

Supplier: Matrix Scientific

076627-500MG 076627-1G
102547-168EA 755.38 USD
102547-168 102547-170
(2S)-1-(3,3-Dimethyl-2-oxopentanoyl)pyrrolidine-2-carboxylic acid ≥97%
(2S)-1-(3,3-Dimethyl-2-oxopentanoyl)pyrrolidine-2-carboxylic acid

Hazard: Irritant

Formula: C₁₂H₁₉NO₄
MW: 241.29 g/mol
Melting Pt: 84…87 °C
MDL Number: MFCD02179274
CAS Number: 186268-78-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR