1,3-Diethyl-2{[(pyridin-3-yl)amino]methylidene}propanedioate ≥90%

Supplier: Matrix Scientific

076626-500MG 076626-1G
102547-164EA 298.46 USD
102547-164 102547-166
1,3-Diethyl-2{[(pyridin-3-yl)amino]methylidene}propanedioate ≥90%
1,3-Diethyl-2{[(pyridin-3-yl)amino]methylidene}propanedioate

Hazard: Irritant

Formula: C₁₃H₁₆N₂O₄
MW: 264.28 g/mol
Melting Pt: 68…70 °C
MDL Number: MFCD00487270
CAS Number: 14029-71-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR