1H,4H,5H,6H-Cyclopenta[c]pyrazole-3-carbonitrile ≥97%

Supplier: Matrix Scientific

076625-500MG 076625-1G 076625-5G
102547-158EA 803.08 USD
102547-158 102547-160 102547-162
1H,4H,5H,6H-Cyclopenta[c]pyrazole-3-carbonitrile ≥97%
1H,4H,5H,6H-Cyclopenta[c]pyrazole-3-carbonitrile

Hazard: Irritant

Formula: C₇H₇N₃
Melting Pt: 179…182 °C
MDL Number: MFCD11109619
CAS Number: 851776-29-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR